The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002379.3 | NP_002370.1 | 1 - 2 | 122 | 126 |
GenBank mRNA | AK075519.1 | - | 1 - 2 | 125 | 126 |
AK312949.1 | - | 1 - 2 | 122 | 126 |