Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKHD1
Entrez Gene ID
5314
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Aliases
ARPKD, FCYT, TIGM1
Chromosome Location
Chr.6: 51613291 - 52087625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138694.3 NP_619639.3 58 - 59 10103 96
NM_170724.2 NP_733842.2 58 - 59 10103 96
XM_011514680.2 XP_011512982.1 58 - 59 10447 96
XM_011514682.2 XP_011512984.1 57 - 58 9931 96
XM_011514683.2 XP_011512985.1 59 - 60 9427 96
XM_011514684.2 XP_011512986.1 50 - 51 9213 96
XM_011514685.1 XP_011512987.1 58 - 59 10103 96
XM_011514686.2 XP_011512988.1 58 - 59 10103 96
XM_011514687.1 XP_011512989.1 58 - 59 10103 96
XM_011514690.2 XP_011512992.1 23 - 24 3989 96
XM_011514691.2 XP_011512993.1 23 - 24 10014 96
XM_017010944.1 XP_016866433.1 58 - 59 9929 96
XM_017010945.1 XP_016866434.1 57 - 58 9994 96
XM_017010946.1 XP_016866435.1 56 - 57 9874 96
XM_017010947.1 XP_016866436.1 56 - 57 9805 96
XM_017010948.1 XP_016866437.1 49 - 50 9887 96
XM_017010949.1 XP_016866438.1 40 - 41 9066 96
XM_017010950.1 XP_016866439.1 58 - 59 10103 96
GenBank mRNA AF480064.1 - 58 - 59 10049 96
AK128031.1 - 22 - 23 4026 96
AY074797.1 - 58 - 59 10103 96
AY092083.1 - 58 - 59 10089 96
BX647896.1 - 6 - 7 1437 96

View Full Product Details