Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX13
Entrez Gene ID
5194
Gene Name
peroxisomal biogenesis factor 13
Gene Aliases
NALD, PBD11A, PBD11B, ZWS
Chromosome Location
Chr.2: 61017677 - 61051990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC025779.1 - 1 - 2 547 107

View Full Product Details