Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCSK5
Entrez Gene ID
5125
Gene Name
proprotein convertase subtilisin/kexin type 5
Gene Aliases
PC5, PC6, PC6A, SPC6
Chromosome Location
Chr.9: 75890601 - 76362339 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190482.1 NP_001177411.1 15 - 16 2548 102
NM_006200.5 NP_006191.2 15 - 16 2548 102
NR_120409.1 14 - 15 2444 102
XM_005252039.3 XP_005252096.1 15 - 16 2591 102
XM_011518769.2 XP_011517071.1 15 - 16 2591 102
XM_011518770.2 XP_011517072.1 6 - 7 5364 102
XM_017014800.1 XP_016870289.1 7 - 8 864 102
GenBank mRNA AK225773.1 - 14 - 15 2441 102
AL834522.1 - 15 - 16 2078 102
BC012064.1 - 15 - 16 2487 102
U49114.1 - 15 - 16 1968 102
U56387.2 - 15 - 16 2022 102

View Full Product Details