The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001323619.1 | NP_001310548.1 | 1 - 2 | 104 | 102 |
NM_001323620.1 | NP_001310549.1 | 1 - 2 | 494 | 102 | |
NM_018671.4 | NP_061141.2 | 1 - 2 | 494 | 102 | |
XM_011521779.2 | XP_011520081.1 | 4 - 5 | 1090 | 102 | |
GenBank mRNA | AB014736.1 | - | 1 - 2 | 78 | 102 |
AK000905.1 | - | 1 - 2 | 70 | 102 | |
AK293538.1 | - | 1 - 2 | 70 | 102 | |
AK302979.1 | - | 1 - 2 | 111 | 102 | |
BC006214.1 | - | 1 - 2 | 77 | 102 | |
BC037992.1 | - | 1 - 2 | 77 | 102 |