The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039675.1 | NP_001034764.1 | 5 - 6 | 1004 | 83 |
NM_001323619.1 | NP_001310548.1 | 2 - 3 | 263 | 83 | |
NM_001323620.1 | NP_001310549.1 | 2 - 3 | 653 | 83 | |
NM_001323621.1 | NP_001310550.1 | 2 - 3 | 608 | 83 | |
NM_018671.4 | NP_061141.2 | 2 - 3 | 653 | 83 | |
XM_005254963.3 | XP_005255020.1 | 2 - 3 | 225 | 83 | |
XM_011521779.2 | XP_011520081.1 | 5 - 6 | 1249 | 83 | |
XM_011521780.2 | XP_011520082.1 | 1 - 2 | 155 | 83 | |
XM_017022407.1 | XP_016877896.1 | 2 - 3 | 225 | 83 | |
GenBank mRNA | AB014729.1 | - | 2 - 3 | 195 | 83 |
AB014736.1 | - | 2 - 3 | 237 | 83 | |
AK000905.1 | - | 2 - 3 | 229 | 83 | |
AK125721.1 | - | 5 - 6 | 1004 | 83 | |
AK291622.1 | - | 2 - 3 | 208 | 83 | |
AK293538.1 | - | 2 - 3 | 229 | 83 | |
AK302979.1 | - | 2 - 3 | 270 | 83 | |
BC006214.1 | - | 2 - 3 | 236 | 83 | |
BC037992.1 | - | 2 - 3 | 236 | 83 | |
BC045635.1 | - | 5 - 6 | 1004 | 83 |