The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039675.1 | NP_001034764.1 | 6 - 7 | 1035 | 68 |
NM_001323619.1 | NP_001310548.1 | 3 - 4 | 294 | 68 | |
NM_001323621.1 | NP_001310550.1 | 3 - 4 | 639 | 68 | |
NM_018671.4 | NP_061141.2 | 3 - 4 | 684 | 68 | |
XM_011521779.2 | XP_011520081.1 | 6 - 7 | 1280 | 68 | |
XM_011521780.2 | XP_011520082.1 | 2 - 3 | 186 | 68 | |
GenBank mRNA | AB014729.1 | - | 3 - 4 | 226 | 68 |
AB014736.1 | - | 3 - 4 | 268 | 68 | |
AK125721.1 | - | 6 - 7 | 1035 | 68 | |
AK291622.1 | - | 3 - 4 | 239 | 68 | |
AK302979.1 | - | 3 - 4 | 301 | 68 | |
BC006214.1 | - | 3 - 4 | 267 | 68 | |
BC037992.1 | - | 3 - 4 | 267 | 68 | |
BC045635.1 | - | 6 - 7 | 1035 | 68 |