The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039675.1 | NP_001034764.1 | 8 - 9 | 1314 | 81 |
NM_001323619.1 | NP_001310548.1 | 5 - 6 | 573 | 81 | |
NM_001323620.1 | NP_001310549.1 | 5 - 6 | 1111 | 81 | |
NM_001323621.1 | NP_001310550.1 | 5 - 6 | 918 | 81 | |
NM_018671.4 | NP_061141.2 | 5 - 6 | 963 | 81 | |
XM_005254963.3 | XP_005255020.1 | 5 - 6 | 683 | 81 | |
XM_011521779.2 | XP_011520081.1 | 8 - 9 | 1559 | 81 | |
XM_011521780.2 | XP_011520082.1 | 4 - 5 | 465 | 81 | |
XM_017022407.1 | XP_016877896.1 | 5 - 6 | 683 | 81 | |
GenBank mRNA | AB014729.1 | - | 5 - 6 | 505 | 81 |
AB014736.1 | - | 5 - 6 | 547 | 81 | |
AK000905.1 | - | 4 - 5 | 841 | 81 | |
AK125721.1 | - | 8 - 9 | 1314 | 81 | |
AK291622.1 | - | 5 - 6 | 518 | 81 | |
AK293538.1 | - | 5 - 6 | 687 | 81 | |
AK296964.1 | - | 5 - 6 | 540 | 81 | |
AK302979.1 | - | 5 - 6 | 580 | 81 | |
BC006214.1 | - | 5 - 6 | 546 | 81 | |
BC037992.1 | - | 5 - 6 | 546 | 81 | |
BC045635.1 | - | 8 - 9 | 1314 | 81 |