The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000808.3 | NP_000799.1 | 3 - 4 | 499 | 77 |
XM_006724811.2 | XP_006724874.1 | 3 - 4 | 499 | 77 | |
GenBank mRNA | A28102.1 | - | 3 - 4 | 345 | 77 |
AK312404.1 | - | 3 - 4 | 477 | 77 | |
BC028629.1 | - | 3 - 4 | 498 | 77 | |
DA402645.1 | - | 3 - 4 | 499 | 77 | |
KJ620007.1 | - | 9 - 10 | 1128 | 77 | |
S62908.1 | - | 3 - 4 | 344 | 77 |