The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301065.1 | NP_001287994.1 | 9 - 10 | 1050 | 73 |
NM_007180.2 | NP_009111.2 | 10 - 11 | 1143 | 73 | |
GenBank mRNA | AB000824.1 | - | 10 - 11 | 1153 | 73 |
AK091981.1 | - | 8 - 9 | 1012 | 73 | |
AK223140.1 | - | 10 - 11 | 1117 | 73 | |
AK223143.1 | - | 10 - 11 | 1121 | 73 | |
BC109206.2 | - | 9 - 10 | 1044 | 73 |