Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF638
Entrez Gene ID
27332
Gene Name
zinc finger protein 638
Gene Aliases
NP220, ZFML, Zfp638
Chromosome Location
Chr.2: 71276543 - 71435061 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014972.2 NP_001014972.1 12 - 13 2810 149
NM_001252612.1 NP_001239541.1 12 - 13 3136 149
NM_001252613.1 NP_001239542.1 12 - 13 2820 149
NM_014497.4 NP_055312.2 12 - 13 2820 149
XM_005264263.1 XP_005264320.1 11 - 12 1284 149
XM_006711989.1 XP_006712052.1 11 - 12 1296 149
XM_011532768.2 XP_011531070.2 12 - 13 2906 149
XM_017003809.1 XP_016859298.1 12 - 13 2911 149
XM_017003810.1 XP_016859299.1 12 - 13 2821 149
XM_017003811.1 XP_016859300.1 12 - 13 2906 149
GenBank mRNA AF273049.1 - 2 - 3 253 149
AF534078.1 - 11 - 12 2500 149
AK291198.1 - 12 - 13 2784 149
AK303723.1 - 10 - 11 1280 149
BC083513.1 - 12 - 13 2779 149
BC143728.1 - 12 - 13 2773 149
BX537425.1 - 12 - 13 2784 149
CR749322.1 - 12 - 13 3133 149

View Full Product Details