The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014972.2 | NP_001014972.1 | 15 - 16 | 3008 | 78 |
NM_001252612.1 | NP_001239541.1 | 15 - 16 | 3334 | 78 | |
NM_001252613.1 | NP_001239542.1 | 15 - 16 | 3018 | 78 | |
NM_014497.4 | NP_055312.2 | 15 - 16 | 3018 | 78 | |
XM_005264263.1 | XP_005264320.1 | 14 - 15 | 1482 | 78 | |
XM_006711989.1 | XP_006712052.1 | 14 - 15 | 1494 | 78 | |
XM_011532768.2 | XP_011531070.2 | 15 - 16 | 3104 | 78 | |
XM_017003809.1 | XP_016859298.1 | 15 - 16 | 3109 | 78 | |
XM_017003810.1 | XP_016859299.1 | 15 - 16 | 3019 | 78 | |
XM_017003811.1 | XP_016859300.1 | 15 - 16 | 3104 | 78 | |
GenBank mRNA | AF273049.1 | - | 5 - 6 | 451 | 78 |
AF534078.1 | - | 14 - 15 | 2698 | 78 | |
AK291198.1 | - | 15 - 16 | 2982 | 78 | |
AK303723.1 | - | 13 - 14 | 1478 | 78 | |
BC083513.1 | - | 15 - 16 | 2977 | 78 | |
BC143728.1 | - | 15 - 16 | 2971 | 78 | |
BX537425.1 | - | 15 - 16 | 2982 | 78 | |
CR749322.1 | - | 15 - 16 | 3331 | 78 | |
D83032.1 | - | 15 - 16 | 3010 | 78 |