Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF638
Entrez Gene ID
27332
Gene Name
zinc finger protein 638
Gene Aliases
NP220, ZFML, Zfp638
Chromosome Location
Chr.2: 71276543 - 71435061 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014972.2 NP_001014972.1 23 - 24 4905 142
NM_001252612.1 NP_001239541.1 23 - 24 5231 142
NM_001252613.1 NP_001239542.1 23 - 24 4915 142
NM_014497.4 NP_055312.2 23 - 24 4915 142
XM_005264263.1 XP_005264320.1 22 - 23 3379 142
XM_006711989.1 XP_006712052.1 22 - 23 3391 142
XM_011532768.2 XP_011531070.2 22 - 23 4875 142
XM_017003809.1 XP_016859298.1 23 - 24 5006 142
XM_017003810.1 XP_016859299.1 22 - 23 4790 142
GenBank mRNA AF273049.1 - 13 - 14 2348 142
AF534078.1 - 21 - 22 3370 142
AK093578.1 - 2 - 3 486 142
AK128706.1 - 4 - 5 2872 142
AK291198.1 - 23 - 24 4879 142
BC083513.1 - 23 - 24 4874 142
BC143728.1 - 23 - 24 4868 142
BX537425.1 - 23 - 24 4879 142
CR749322.1 - 23 - 24 5228 142
D83032.1 - 23 - 24 4907 142

View Full Product Details