Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF638
Entrez Gene ID
27332
Gene Name
zinc finger protein 638
Gene Aliases
NP220, ZFML, Zfp638
Chromosome Location
Chr.2: 71276543 - 71435061 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014972.2 NP_001014972.1 24 - 25 5860 118
NM_001252612.1 NP_001239541.1 24 - 25 6186 118
NM_001252613.1 NP_001239542.1 24 - 25 5870 118
NM_014497.4 NP_055312.2 24 - 25 5870 118
XM_005264263.1 XP_005264320.1 23 - 24 4334 118
XM_006711989.1 XP_006712052.1 23 - 24 4346 118
XM_011532768.2 XP_011531070.2 23 - 24 5830 118
XM_017003809.1 XP_016859298.1 24 - 25 5961 118
XM_017003810.1 XP_016859299.1 23 - 24 5745 118
GenBank mRNA AF273049.1 - 14 - 15 3303 118
AK093578.1 - 3 - 4 1441 118
AK128706.1 - 5 - 6 3828 118
BC083513.1 - 24 - 25 5829 118
BC143728.1 - 24 - 25 5823 118
BX537425.1 - 24 - 25 5834 118
CR749322.1 - 24 - 25 6183 118
D83032.1 - 24 - 25 5862 118

View Full Product Details