Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP10
Entrez Gene ID
4319
Gene Name
matrix metallopeptidase 10
Gene Aliases
SL-2, STMY2
Chromosome Location
Chr.11: 102770502 - 102780628 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002425.2 NP_002416.1 6 - 7 958 83
GenBank mRNA AK222601.1 - 6 - 7 958 83
AK313960.1 - 6 - 7 958 83
BC002591.2 - 6 - 7 945 83
BT007442.1 - 6 - 7 921 83
X07820.1 - 6 - 7 943 83

View Full Product Details