The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015500.1 | NP_056315.1 | 9 - 10 | 1384 | 105 |
NM_199050.2 | NP_950251.1 | 8 - 9 | 883 | 105 | |
XM_005261109.4 | XP_005261166.1 | 8 - 9 | 924 | 105 | |
XM_011529522.1 | XP_011527824.1 | 9 - 10 | 1484 | 105 | |
XM_011529523.2 | XP_011527825.1 | 10 - 11 | 2150 | 105 | |
XM_011529524.2 | XP_011527826.1 | 9 - 10 | 1484 | 105 | |
XM_017028315.1 | XP_016883804.1 | 6 - 7 | 620 | 105 | |
GenBank mRNA | AB047784.1 | - | 9 - 10 | 1384 | 105 |
AB047785.1 | - | 9 - 10 | 1384 | 105 | |
AK128550.1 | - | 6 - 7 | 723 | 105 | |
AL833323.1 | - | 8 - 9 | 883 | 105 | |
BC062323.1 | - | 8 - 9 | 865 | 105 | |
CR627455.1 | - | 1 - 2 | 1158 | 105 |