The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291746.1 | NP_001278675.1 | 4 - 5 | 735 | 152 |
NM_002908.3 | NP_002899.1 | 4 - 5 | 735 | 152 | |
XM_011533010.2 | XP_011531312.1 | 2 - 3 | 222 | 152 | |
XM_017004627.1 | XP_016860116.1 | 4 - 5 | 656 | 152 | |
GenBank mRNA | AK303926.1 | - | 4 - 5 | 656 | 152 |
BC117191.1 | - | 4 - 5 | 574 | 152 | |
BC143885.1 | - | 4 - 5 | 576 | 152 | |
X75042.1 | - | 4 - 5 | 566 | 152 |