The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201428.1 | NP_001188357.1 | 1 - 2 | 124 | 69 |
GenBank mRNA | AL117430.1 | - | 1 - 2 | 90 | 69 |