Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHRF1
Entrez Gene ID
57661
Gene Name
PHD and ring finger domains 1
Gene Aliases
PPP1R125, RNF221
Chromosome Location
Chr.11: 576446 - 612225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286581.1 NP_001273510.1 10 - 11 1322 76
NM_001286582.1 NP_001273511.1 10 - 11 1319 76
NM_001286583.1 NP_001273512.1 10 - 11 1310 76
NM_020901.3 NP_065952.2 10 - 11 1322 76
XM_005253025.4 XP_005253082.1 10 - 11 1280 76
XM_005253027.3 XP_005253084.1 10 - 11 1268 76
XM_011520237.2 XP_011518539.1 9 - 10 1163 76
GenBank mRNA AB040975.1 - 10 - 11 1282 76
BC112931.1 - 8 - 9 3067 76
BC136615.1 - 10 - 11 1247 76
BC144293.1 - 10 - 11 1247 76
BC144294.1 - 10 - 11 1235 76
BC144295.1 - 10 - 11 1244 76
BC146771.1 - 10 - 11 1282 76

View Full Product Details