The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004922.3 | NP_004913.2 | 15 - 16 | 2194 | 70 |
NM_198597.2 | NP_940999.1 | 14 - 15 | 2107 | 70 | |
XM_011540380.2 | XP_011538682.1 | 15 - 16 | 2199 | 70 | |
XM_011540381.2 | XP_011538683.1 | 16 - 17 | 2472 | 70 | |
XM_017016967.1 | XP_016872456.1 | 16 - 17 | 2184 | 70 | |
XM_017016968.1 | XP_016872457.1 | 15 - 16 | 2323 | 70 | |
GenBank mRNA | AK098512.1 | - | 2 - 3 | 167 | 70 |
AK299826.1 | - | 14 - 15 | 2013 | 70 | |
AL832110.1 | - | 14 - 15 | 2089 | 70 | |
BC018928.2 | - | 14 - 15 | 2095 | 70 | |
D38555.1 | - | 14 - 15 | 2128 | 70 |