The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020979.4 | NP_066189.3 | 1 - 2 | 198 | 84 |
XM_005276975.4 | XP_005277032.1 | 1 - 2 | 734 | 84 | |
GenBank mRNA | AB000520.1 | - | 1 - 2 | 99 | 84 |
BF434397.1 | - | 1 - 2 | 84 | ||
DN995670.1 | - | 1 - 2 | 153 | 84 |