The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020979.4 | NP_066189.3 | 2 - 3 | 957 | 60 |
XM_005276975.4 | XP_005277032.1 | 2 - 3 | 1493 | 60 | |
XM_005276976.3 | XP_005277033.1 | 2 - 3 | 1485 | 60 | |
XM_005276977.4 | XP_005277034.1 | 2 - 3 | 992 | 60 | |
XM_005276979.3 | XP_005277036.1 | 2 - 3 | 908 | 60 | |
XM_011515740.2 | XP_011514042.1 | 2 - 3 | 1739 | 60 | |
XM_017011680.1 | XP_016867169.1 | 2 - 3 | 993 | 60 | |
XM_017011681.1 | XP_016867170.1 | 2 - 3 | 911 | 60 | |
GenBank mRNA | AB000520.1 | - | 2 - 3 | 858 | 60 |