The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001916.4 | NP_001907.2 | 5 - 6 | 840 | 73 |
XM_017013102.1 | XP_016868591.1 | 4 - 5 | 1373 | 73 | |
GenBank mRNA | AK026633.1 | - | 5 - 6 | 798 | 73 |
AK225796.1 | - | 4 - 5 | 1441 | 73 | |
BC001006.1 | - | 5 - 6 | 786 | 73 | |
BC015616.2 | - | 5 - 6 | 780 | 73 | |
BC020566.1 | - | 5 - 6 | 779 | 73 | |
BC024650.1 | - | 5 - 6 | 747 | 73 | |
BQ574597.1 | - | 2 - 3 | 73 | ||
BT019798.1 | - | 5 - 6 | 775 | 73 | |
CR541674.1 | - | 5 - 6 | 775 | 73 | |
M16597.1 | - | 4 - 5 | 481 | 73 | |
X06994.1 | - | 5 - 6 | 783 | 73 |