The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016377.3 | NP_057461.2 | 7 - 8 | 929 | 121 |
GenBank mRNA | AF152929.1 | - | 6 - 7 | 796 | 121 |
AK300587.1 | - | 7 - 8 | 884 | 121 |