Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIN1
Entrez Gene ID
9610
Gene Name
Ras and Rab interactor 1
Gene Aliases
-
Chromosome Location
Chr.11: 66332063 - 66337235 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004292.2 NP_004283.2 2 - 3 401 71
XM_011545399.2 XP_011543701.1 2 - 3 750 71
XM_011545400.2 XP_011543702.1 2 - 3 306 71
XM_011545401.2 XP_011543703.1 2 - 3 275 71
XM_017018587.1 XP_016874076.1 2 - 3 306 71
GenBank mRNA BC014417.1 - 2 - 3 291 71
KU178481.1 - 2 - 3 274 71
KU178482.1 - 2 - 3 274 71
L36463.1 - 2 - 3 401 71

View Full Product Details