The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184854.1 | NP_001171783.1 | 4 - 5 | 531 | 71 |
NM_017566.3 | NP_060036.2 | 6 - 7 | 702 | 71 | |
XM_005255994.3 | XP_005256051.1 | 6 - 7 | 707 | 71 | |
XM_005255999.1 | XP_005256056.1 | 5 - 6 | 622 | 71 | |
XM_006721201.1 | XP_006721264.1 | 6 - 7 | 722 | 71 | |
XM_006721202.1 | XP_006721265.1 | 6 - 7 | 722 | 71 | |
XM_006721203.2 | XP_006721266.1 | 6 - 7 | 722 | 71 | |
XM_006721204.3 | XP_006721267.1 | 6 - 7 | 722 | 71 | |
XM_017023344.1 | XP_016878833.1 | 6 - 7 | 707 | 71 | |
XM_017023345.1 | XP_016878834.1 | 6 - 7 | 647 | 71 | |
GenBank mRNA | AK023986.1 | - | 6 - 7 | 659 | 71 |
AK097097.1 | - | 4 - 5 | 1583 | 71 | |
AK098630.1 | - | 2 - 3 | 150 | 71 | |
AL133107.1 | - | 4 - 5 | 465 | 71 | |
BC001044.2 | - | 4 - 5 | 438 | 71 | |
BC022969.1 | - | 6 - 7 | 646 | 71 |