The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006076.4 | NP_006067.3 | 1 - 2 | 361 | 82 |
XM_005250306.2 | XP_005250363.1 | 1 - 2 | 347 | 82 | |
XM_017012113.1 | XP_016867602.1 | 1 - 2 | 347 | 82 | |
XM_017012114.1 | XP_016867603.1 | 1 - 2 | 347 | 82 | |
GenBank mRNA | AF015042.1 | - | 1 - 2 | 285 | 82 |
AK314954.1 | - | 1 - 2 | 347 | 82 | |
BC009393.2 | - | 1 - 2 | 360 | 82 | |
BC017329.2 | - | 1 - 2 | 313 | 82 | |
BF310674.1 | - | 1 - 2 | 359 | 81 | |
BF314987.1 | - | 1 - 2 | 361 | 82 |