Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGFG2
Entrez Gene ID
3268
Gene Name
ArfGAP with FG repeats 2
Gene Aliases
HRBL, RABR
Chromosome Location
Chr.7: 100539211 - 100568220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006076.4 NP_006067.3 5 - 6 891 81
XM_005250306.2 XP_005250363.1 5 - 6 877 81
XM_017012112.1 XP_016867601.1 5 - 6 832 81
XM_017012113.1 XP_016867602.1 5 - 6 877 81
GenBank mRNA AF015042.1 - 5 - 6 815 81
AK314954.1 - 5 - 6 877 81
BC017329.2 - 5 - 6 843 81

View Full Product Details