Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZAN
Entrez Gene ID
7455
Gene Name
zonadhesin (gene/pseudogene)
Gene Aliases
-
Chromosome Location
Chr.7: 100733599 - 100797797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003386.2 NP_003377.2 20 - 21 4148 94
NM_173059.2 NP_775082.2 20 - 21 4148 94
NR_111917.1 20 - 21 4148 94
NR_111918.1 20 - 21 4148 94
NR_111919.1 20 - 21 4148 94
XM_011516555.2 XP_011514857.1 20 - 21 3881 94
XM_017012581.1 XP_016868070.1 7 - 8 1637 94
GenBank mRNA AF332975.1 - 20 - 21 4148 94
AF332976.1 - 20 - 21 4148 94
AF332977.1 - 20 - 21 4148 94
AF332978.1 - 20 - 21 4148 94
AF332979.1 - 20 - 21 4148 94
AF332980.1 - 20 - 21 4148 94

View Full Product Details