The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003386.2 | NP_003377.2 | 46 - 47 | 8534 | 69 |
NM_173059.2 | NP_775082.2 | 44 - 45 | 8261 | 69 | |
NR_111917.1 | 46 - 47 | 8599 | 69 | ||
NR_111918.1 | 45 - 46 | 8400 | 69 | ||
NR_111919.1 | 45 - 46 | 8335 | 69 | ||
XM_011516555.2 | XP_011514857.1 | 44 - 45 | 7997 | 69 | |
GenBank mRNA | AF332975.1 | - | 45 - 46 | 8700 | 69 |
AF332976.1 | - | 46 - 47 | 8599 | 69 | |
AF332977.1 | - | 46 - 47 | 8534 | 69 | |
AF332978.1 | - | 45 - 46 | 8400 | 69 | |
AF332979.1 | - | 45 - 46 | 8335 | 69 | |
AF332980.1 | - | 44 - 45 | 8261 | 69 | |
AK302596.1 | - | 23 - 24 | 3550 | 69 |