Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZAN
Entrez Gene ID
7455
Gene Name
zonadhesin (gene/pseudogene)
Gene Aliases
-
Chromosome Location
Chr.7: 100733599 - 100797797 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003386.2 NP_003377.2 47 - 48 8575 67
NM_173059.2 NP_775082.2 45 - 46 8302 67
NR_111917.1 47 - 48 8640 67
NR_111918.1 46 - 47 8441 67
NR_111919.1 46 - 47 8376 67
XM_011516555.2 XP_011514857.1 45 - 46 8038 67
GenBank mRNA AF332975.1 - 46 - 47 8741 67
AF332976.1 - 47 - 48 8640 67
AF332977.1 - 47 - 48 8575 67
AF332978.1 - 46 - 47 8441 67
AF332979.1 - 46 - 47 8376 67
AF332980.1 - 45 - 46 8302 67
AK302596.1 - 24 - 25 3591 67

View Full Product Details