The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003386.2 | NP_003377.2 | 6 - 7 | 782 | 96 |
NM_173059.2 | NP_775082.2 | 6 - 7 | 782 | 96 | |
NR_111917.1 | 6 - 7 | 782 | 96 | ||
NR_111918.1 | 6 - 7 | 782 | 96 | ||
NR_111919.1 | 6 - 7 | 782 | 96 | ||
XM_011516555.2 | XP_011514857.1 | 5 - 6 | 1187 | 96 | |
GenBank mRNA | AF332975.1 | - | 6 - 7 | 782 | 96 |
AF332976.1 | - | 6 - 7 | 782 | 96 | |
AF332977.1 | - | 6 - 7 | 782 | 96 | |
AF332978.1 | - | 6 - 7 | 782 | 96 | |
AF332979.1 | - | 6 - 7 | 782 | 96 | |
AF332980.1 | - | 6 - 7 | 782 | 96 |