Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAMK2G
Entrez Gene ID
818
Gene Name
calcium/calmodulin dependent protein kinase II gamma
Gene Aliases
CAMK, CAMK-II, CAMKG
Chromosome Location
Chr.10: 73812501 - 73874591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204492.1 NP_001191421.1 3 - 4 352 83
NM_001222.3 NP_001213.2 3 - 4 352 83
NM_001320898.1 NP_001307827.1 3 - 4 352 83
NM_172169.2 NP_751909.1 3 - 4 352 83
NM_172170.4 NP_751910.1 3 - 4 352 83
NM_172171.2 NP_751911.1 3 - 4 352 83
NM_172173.2 NP_751913.1 3 - 4 352 83
XM_005270195.1 XP_005270252.1 3 - 4 343 83
XM_005270197.1 XP_005270254.1 3 - 4 343 83
XM_005270198.1 XP_005270255.1 3 - 4 343 83
XM_005270199.1 XP_005270256.1 3 - 4 343 83
XM_005270200.1 XP_005270257.1 3 - 4 343 83
XM_005270201.1 XP_005270258.1 3 - 4 343 83
XM_005270203.1 XP_005270260.1 3 - 4 343 83
XM_005270205.1 XP_005270262.1 3 - 4 343 83
XM_006717993.1 XP_006718056.1 3 - 4 343 83
XM_006717996.1 XP_006718059.1 3 - 4 343 83
XM_006717997.1 XP_006718060.1 3 - 4 343 83
XM_006717998.1 XP_006718061.1 3 - 4 343 83
XM_011540225.2 XP_011538527.1 3 - 4 291 83
XM_017016716.1 XP_016872205.1 3 - 4 343 83
XM_017016717.1 XP_016872206.1 3 - 4 291 83
XM_017016718.1 XP_016872207.1 3 - 4 343 83
XM_017016719.1 XP_016872208.1 3 - 4 343 83
XM_017016720.1 XP_016872209.1 3 - 4 343 83
XM_017016721.1 XP_016872210.1 3 - 4 418 83
XM_017016722.1 XP_016872211.1 3 - 4 343 83
XM_017016723.1 XP_016872212.1 3 - 4 343 83
XM_017016724.1 XP_016872213.1 3 - 4 343 83
XM_017016725.1 XP_016872214.1 3 - 4 343 83
XM_017016726.1 XP_016872215.1 3 - 4 343 83
XM_017016727.1 XP_016872216.1 3 - 4 343 83
XM_017016728.1 XP_016872217.1 3 - 4 291 83
XM_017016730.1 XP_016872219.1 3 - 4 343 83
XM_017016733.1 XP_016872222.1 3 - 4 291 83
XM_017016735.1 XP_016872224.1 3 - 4 343 83
XM_017016736.1 XP_016872225.1 3 - 4 418 83
XM_017016738.1 XP_016872227.1 3 - 4 304 83
XM_017016739.1 XP_016872228.1 3 - 4 475 83
GenBank mRNA AK131199.1 - 3 - 4 275 83
AK225771.1 - 3 - 4 352 83
AK291704.1 - 3 - 4 344 83
BC034044.1 - 3 - 4 343 83
BU165166.1 - 3 - 4 349 83
DR002605.1 - 3 - 4 359 83
L07044.1 - 3 - 4 243 83
U66064.1 - 3 - 4 275 83

View Full Product Details