The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163435.2 | NP_001156907.1 | 14 - 15 | 1712 | 105 |
NM_001163436.2 | NP_001156908.1 | 14 - 15 | 1533 | 105 | |
NM_001163437.2 | NP_001156909.1 | 14 - 15 | 1595 | 105 | |
NM_001290768.1 | NP_001277697.1 | 15 - 16 | 1813 | 105 | |
NM_033115.4 | NP_149106.2 | 12 - 13 | 1523 | 105 | |
XM_011532417.2 | XP_011530719.1 | 14 - 15 | 1632 | 105 | |
XM_017008846.1 | XP_016864335.1 | 14 - 15 | 1716 | 105 | |
XM_017008847.1 | XP_016864336.1 | 13 - 14 | 1376 | 105 | |
XM_017008848.1 | XP_016864337.1 | 14 - 15 | 1388 | 105 | |
XM_017008849.1 | XP_016864338.1 | 12 - 13 | 1199 | 105 | |
GenBank mRNA | AB449876.1 | - | 13 - 14 | 1347 | 105 |
AF161420.1 | - | 2 - 3 | 186 | 105 | |
AK074305.1 | - | 14 - 15 | 1467 | 105 | |
AK098157.1 | - | 5 - 6 | 1000 | 105 | |
AK315571.1 | - | 14 - 15 | 1655 | 105 | |
BC009208.2 | - | 12 - 13 | 1485 | 105 | |
BC068496.1 | - | 14 - 15 | 1536 | 105 | |
BX647851.1 | - | 16 - 17 | 1757 | 105 |