Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 20 - 21 2007 101
NM_001324418.1 NP_001311347.1 20 - 21 2010 101
NM_001324419.1 NP_001311348.1 20 - 21 2007 101
NM_001324420.1 NP_001311349.1 20 - 21 2010 101
NM_001324421.1 NP_001311350.1 20 - 21 2010 101
NM_004194.4 NP_004185.1 20 - 21 2010 101
NM_016351.5 NP_057435.2 20 - 21 2010 101
NM_021721.4 NP_068367.1 20 - 21 2010 101
NM_021722.5 NP_068368.2 20 - 21 2010 101
NM_021723.4 NP_068369.1 20 - 21 2010 101
XM_005250445.3 XP_005250502.1 19 - 20 1858 101
XM_006716028.3 XP_006716091.1 19 - 20 1858 101
XM_006716029.3 XP_006716092.1 19 - 20 1858 101
XM_011516318.1 XP_011514620.1 20 - 21 1902 101
XM_011516319.2 XP_011514621.1 20 - 21 1986 101
XM_011516320.2 XP_011514622.1 19 - 20 1858 101
XM_011516321.2 XP_011514623.1 19 - 20 1858 101
XM_011516322.1 XP_011514624.1 20 - 21 1902 101
XM_011516323.1 XP_011514625.1 20 - 21 1902 101
XM_011516324.1 XP_011514626.1 20 - 21 1574 101
XM_017012329.1 XP_016867818.1 21 - 22 1953 101
XM_017012330.1 XP_016867819.1 21 - 22 1953 101
XM_017012331.1 XP_016867820.1 20 - 21 1983 101
XM_017012332.1 XP_016867821.1 20 - 21 1902 101
XM_017012333.1 XP_016867822.1 20 - 21 1902 101
XM_017012334.1 XP_016867823.1 21 - 22 1953 101
XM_017012335.1 XP_016867824.1 21 - 22 1953 101
XM_017012336.1 XP_016867825.1 20 - 21 1902 101
XM_017012337.1 XP_016867826.1 21 - 22 1953 101
XM_017012338.1 XP_016867827.1 20 - 21 1981 101
XM_017012339.1 XP_016867828.1 21 - 22 1953 101
XM_017012340.1 XP_016867829.1 9 - 10 633 101
XM_017012341.1 XP_016867830.1 19 - 20 1630 101
GenBank mRNA AB009671.1 - 20 - 21 1808 101
AF073291.2 - 20 - 21 1733 101
AF155381.1 - 20 - 21 1733 101
AF155382.1 - 20 - 21 1733 101
AF158637.1 - 19 - 20 1570 101
AK125432.1 - 19 - 20 1873 101
AK291010.1 - 20 - 21 1857 101
BC125118.1 - 20 - 21 1764 101

View Full Product Details