Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAM22
Entrez Gene ID
53616
Gene Name
ADAM metallopeptidase domain 22
Gene Aliases
ADAM 22, MDC2
Chromosome Location
Chr.7: 87934143 - 88202889 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001324417.1 NP_001311346.1 29 - 30 2909 146
NM_001324418.1 NP_001311347.1 31 - 32 3194 146
NM_001324419.1 NP_001311348.1 31 - 32 3191 146
NM_001324420.1 NP_001311349.1 30 - 31 3107 146
NM_016351.5 NP_057435.2 29 - 30 2912 146
NM_021722.5 NP_068368.2 30 - 31 2999 146
NM_021723.4 NP_068369.1 30 - 31 3020 146
XM_005250445.3 XP_005250502.1 29 - 30 2868 146
XM_006716028.3 XP_006716091.1 31 - 32 3066 146
XM_011516318.1 XP_011514620.1 32 - 33 3197 146
XM_011516319.2 XP_011514621.1 32 - 33 3281 146
XM_011516320.2 XP_011514622.1 30 - 31 2979 146
XM_011516321.2 XP_011514623.1 30 - 31 2955 146
XM_011516322.1 XP_011514624.1 32 - 33 3110 146
XM_011516323.1 XP_011514625.1 31 - 32 3002 146
XM_011516324.1 XP_011514626.1 32 - 33 2869 146
XM_017012329.1 XP_016867818.1 32 - 33 3053 146
XM_017012330.1 XP_016867819.1 31 - 32 3050 146
XM_017012331.1 XP_016867820.1 31 - 32 3083 146
XM_017012332.1 XP_016867821.1 31 - 32 2999 146
XM_017012334.1 XP_016867823.1 31 - 32 2966 146
XM_017012335.1 XP_016867824.1 31 - 32 2942 146
XM_017012336.1 XP_016867825.1 30 - 31 2915 146
XM_017012338.1 XP_016867827.1 30 - 31 2991 146
XM_017012339.1 XP_016867828.1 30 - 31 2855 146
XM_017012340.1 XP_016867829.1 21 - 22 1928 146
XM_017012341.1 XP_016867830.1 28 - 29 2532 146
GenBank mRNA AF073291.2 - 29 - 30 2635 146
AF155382.1 - 30 - 31 2743 146
AF158637.1 - 29 - 30 2580 146
AK291010.1 - 29 - 30 2759 146
BC125118.1 - 29 - 30 2666 146
CA429662.1 - 1 - 2 146

View Full Product Details