Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPSS1
Entrez Gene ID
9061
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 1
Gene Aliases
ATPSK1, PAPSS, SK1
Chromosome Location
Chr.4: 107613666 - 107720263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005443.4 NP_005434.4 2 - 3 257 67
XM_011532400.1 XP_011530702.1 2 - 3 237 67
XM_011532401.1 XP_011530703.1 2 - 3 323 67
GenBank mRNA AF016496.1 - 2 - 3 173 67
AF033026.1 - 2 - 3 199 67
AF105227.1 - 2 - 3 227 67
AK292774.1 - 2 - 3 207 67
AK297981.1 - 2 - 3 207 67
AU132232.1 - 2 - 3 257 67
BC011392.1 - 2 - 3 130 67
BC032513.1 - 2 - 3 226 67
BC040940.1 - 2 - 3 185 67
BC050627.1 - 2 - 3 193 67
BQ437924.1 - 2 - 3 185 67
CR457028.1 - 2 - 3 173 67
Y10387.1 - 2 - 3 209 67

View Full Product Details