Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC24B
Entrez Gene ID
10427
Gene Name
SEC24 homolog B, COPII coat complex component
Gene Aliases
SEC24
Chromosome Location
Chr.4: 109433715 - 109540459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042734.3 NP_001036199.1 12 - 13 2344 110
NM_001300813.2 NP_001287742.1 14 - 15 2539 110
NM_001318085.1 NP_001305014.1 13 - 14 2446 110
NM_001318086.1 NP_001305015.1 12 - 13 2341 110
NM_006323.4 NP_006314.2 13 - 14 2449 110
XM_005262688.1 XP_005262745.1 14 - 15 2418 110
XM_005262691.1 XP_005262748.1 13 - 14 2310 110
XM_011531535.2 XP_011529837.1 13 - 14 2313 110
XM_011531536.2 XP_011529838.1 13 - 14 2522 110
XM_011531537.2 XP_011529839.1 13 - 14 2228 110
XM_011531539.2 XP_011529841.1 12 - 13 2130 110
XM_011531540.2 XP_011529842.1 8 - 9 959 110
XM_017007652.1 XP_016863141.1 11 - 12 2063 110
GenBank mRNA AJ131245.1 - 13 - 14 2449 110
AK300278.1 - 12 - 13 2132 110
AK304234.1 - 10 - 11 1189 110
BC040137.1 - 13 - 14 2307 110
BC117135.1 - 12 - 13 2287 110
BC143268.1 - 14 - 15 2482 110
BC143276.1 - 12 - 13 2287 110

View Full Product Details