The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042734.3 | NP_001036199.1 | 19 - 20 | 3441 | 139 |
NM_001300813.2 | NP_001287742.1 | 21 - 22 | 3636 | 139 | |
NM_001318085.1 | NP_001305014.1 | 20 - 21 | 3543 | 139 | |
NM_001318086.1 | NP_001305015.1 | 19 - 20 | 3438 | 139 | |
NM_006323.4 | NP_006314.2 | 20 - 21 | 3546 | 139 | |
XM_005262688.1 | XP_005262745.1 | 21 - 22 | 3515 | 139 | |
XM_005262691.1 | XP_005262748.1 | 20 - 21 | 3407 | 139 | |
XM_011531535.2 | XP_011529837.1 | 20 - 21 | 3410 | 139 | |
XM_011531536.2 | XP_011529838.1 | 20 - 21 | 3619 | 139 | |
XM_011531537.2 | XP_011529839.1 | 20 - 21 | 3325 | 139 | |
XM_011531539.2 | XP_011529841.1 | 19 - 20 | 3227 | 139 | |
XM_011531540.2 | XP_011529842.1 | 15 - 16 | 2056 | 139 | |
XM_017007652.1 | XP_016863141.1 | 18 - 19 | 3160 | 139 | |
GenBank mRNA | AJ131245.1 | - | 20 - 21 | 3546 | 139 |
AK300278.1 | - | 19 - 20 | 3229 | 139 | |
AK304234.1 | - | 17 - 18 | 2286 | 139 | |
BC040137.1 | - | 20 - 21 | 3404 | 139 | |
BC117135.1 | - | 19 - 20 | 3384 | 139 | |
BC143268.1 | - | 21 - 22 | 3579 | 139 | |
BC143276.1 | - | 19 - 20 | 3384 | 139 |