The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042734.3 | NP_001036199.1 | 22 - 23 | 3735 | 95 |
NM_001300813.2 | NP_001287742.1 | 24 - 25 | 3930 | 95 | |
NM_001318085.1 | NP_001305014.1 | 23 - 24 | 3837 | 95 | |
NM_001318086.1 | NP_001305015.1 | 22 - 23 | 3732 | 95 | |
NM_006323.4 | NP_006314.2 | 23 - 24 | 3840 | 95 | |
XM_005262688.1 | XP_005262745.1 | 24 - 25 | 3809 | 95 | |
XM_005262691.1 | XP_005262748.1 | 23 - 24 | 3701 | 95 | |
XM_011531535.2 | XP_011529837.1 | 23 - 24 | 3704 | 95 | |
XM_011531536.2 | XP_011529838.1 | 23 - 24 | 3913 | 95 | |
XM_011531537.2 | XP_011529839.1 | 23 - 24 | 3619 | 95 | |
XM_011531539.2 | XP_011529841.1 | 22 - 23 | 3521 | 95 | |
XM_011531540.2 | XP_011529842.1 | 18 - 19 | 2350 | 95 | |
XM_017007652.1 | XP_016863141.1 | 21 - 22 | 3454 | 95 | |
GenBank mRNA | AJ131245.1 | - | 23 - 24 | 3840 | 95 |
AK304234.1 | - | 20 - 21 | 2580 | 95 | |
BC040137.1 | - | 23 - 24 | 3698 | 95 | |
BC117135.1 | - | 22 - 23 | 3678 | 95 | |
BC143268.1 | - | 24 - 25 | 3873 | 95 | |
BC143276.1 | - | 22 - 23 | 3678 | 95 |