The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042734.3 | NP_001036199.1 | 6 - 7 | 1722 | 95 |
NM_001300813.2 | NP_001287742.1 | 8 - 9 | 1917 | 95 | |
NM_001318085.1 | NP_001305014.1 | 7 - 8 | 1824 | 95 | |
NM_001318086.1 | NP_001305015.1 | 6 - 7 | 1719 | 95 | |
NM_006323.4 | NP_006314.2 | 7 - 8 | 1827 | 95 | |
XM_005262688.1 | XP_005262745.1 | 8 - 9 | 1796 | 95 | |
XM_005262691.1 | XP_005262748.1 | 7 - 8 | 1688 | 95 | |
XM_011531535.2 | XP_011529837.1 | 7 - 8 | 1691 | 95 | |
XM_011531536.2 | XP_011529838.1 | 7 - 8 | 1900 | 95 | |
XM_011531537.2 | XP_011529839.1 | 7 - 8 | 1606 | 95 | |
XM_011531539.2 | XP_011529841.1 | 6 - 7 | 1508 | 95 | |
XM_011531540.2 | XP_011529842.1 | 2 - 3 | 337 | 95 | |
XM_017007652.1 | XP_016863141.1 | 5 - 6 | 1441 | 95 | |
GenBank mRNA | AJ131245.1 | - | 7 - 8 | 1827 | 95 |
AK300278.1 | - | 6 - 7 | 1510 | 95 | |
AK304234.1 | - | 4 - 5 | 567 | 95 | |
BC040137.1 | - | 7 - 8 | 1685 | 95 | |
BC117135.1 | - | 6 - 7 | 1665 | 95 | |
BC143268.1 | - | 8 - 9 | 1860 | 95 | |
BC143276.1 | - | 6 - 7 | 1665 | 95 |