The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182476.2 | NP_872282.1 | 1 - 2 | 243 | 116 |
XM_011536807.1 | XP_011535109.1 | 1 - 2 | 215 | 116 | |
XM_011536810.2 | XP_011535112.1 | 1 - 2 | 215 | 116 | |
XM_017021352.1 | XP_016876841.1 | 1 - 2 | 185 | 116 | |
GenBank mRNA | AK130167.1 | - | 1 - 2 | 200 | 116 |
AK222965.1 | - | 1 - 2 | 185 | 116 | |
AK294370.1 | - | 1 - 2 | 185 | 116 | |
AK295517.1 | - | 1 - 2 | 185 | 116 | |
AK296386.1 | - | 1 - 2 | 185 | 116 | |
AK303304.1 | - | 1 - 2 | 194 | 116 | |
AL521052.3 | - | 1 - 2 | 274 | 116 | |
AL557147.3 | - | 1 - 2 | 235 | 116 | |
BC014181.2 | - | 1 - 2 | 192 | 116 | |
BC014483.1 | - | 1 - 2 | 180 | 116 | |
BX342249.2 | - | 1 - 2 | 244 | 116 |