The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019589.2 | NP_062535.2 | 4 - 5 | 2402 | 101 |
XM_005267860.3 | XP_005267917.1 | 4 - 5 | 2446 | 101 | |
XM_011536966.2 | XP_011535268.1 | 5 - 6 | 1561 | 101 | |
GenBank mRNA | AK090435.1 | - | 2 - 3 | 1158 | 101 |
AL832365.1 | - | 1 - 2 | 937 | 101 | |
L40400.1 | - | 1 - 2 | 81 | 101 |