Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOT2
Entrez Gene ID
89941
Gene Name
ras homolog family member T2
Gene Aliases
ARHT2, C16orf39, MIRO-2, MIRO2, RASL
Chromosome Location
Chr.16: 668083 - 674174 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138769.2 NP_620124.1 13 - 14 1216 83
XM_005255660.2 XP_005255717.1 13 - 14 1170 83
XM_005255661.2 XP_005255718.1 12 - 13 1116 83
XM_005255662.2 XP_005255719.1 12 - 13 1113 83
XM_005255663.2 XP_005255720.1 12 - 13 1068 83
XM_006720970.2 XP_006721033.1 13 - 14 1153 83
XM_006720973.2 XP_006721036.1 12 - 13 1051 83
XM_017023825.1 XP_016879314.1 13 - 14 1170 83
XM_017023826.1 XP_016879315.1 13 - 14 1167 83
XM_017023827.1 XP_016879316.1 12 - 13 1116 83
XM_017023828.1 XP_016879317.1 12 - 13 1113 83
XM_017023829.1 XP_016879318.1 12 - 13 1068 83
XM_017023830.1 XP_016879319.1 11 - 12 1014 83
XM_017023831.1 XP_016879320.1 13 - 14 1153 83
XM_017023832.1 XP_016879321.1 10 - 11 914 83
XM_017023833.1 XP_016879322.1 13 - 14 1150 83
XM_017023834.1 XP_016879323.1 11 - 12 960 83
XM_017023835.1 XP_016879324.1 13 - 14 1165 83
XM_017023836.1 XP_016879325.1 12 - 13 1051 83
XM_017023837.1 XP_016879326.1 13 - 14 1150 83
XM_017023838.1 XP_016879327.1 12 - 13 1031 83
XM_017023839.1 XP_016879328.1 13 - 14 1165 83
XM_017023840.1 XP_016879329.1 12 - 13 1179 83
XM_017023841.1 XP_016879330.1 11 - 12 1257 83
XM_017023842.1 XP_016879331.1 12 - 13 1222 83
GenBank mRNA AJ517413.1 - 13 - 14 1099 83
AK024450.1 - 9 - 10 2646 83
AL834163.1 - 12 - 13 1234 83
AY207375.1 - 13 - 14 1213 83
BC014942.1 - 13 - 14 1166 83

View Full Product Details