The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007195.2 | NP_009126.2 | 6 - 7 | 1043 | 178 |
XM_005258188.3 | XP_005258245.1 | 6 - 7 | 1335 | 178 | |
XM_005258192.4 | XP_005258249.1 | 6 - 7 | 1024 | 178 | |
XM_006722378.2 | XP_006722441.1 | 6 - 7 | 1024 | 178 | |
XM_011525796.2 | XP_011524098.1 | 6 - 7 | 1032 | 178 | |
XM_011525797.1 | XP_011524099.1 | 6 - 7 | 1024 | 178 | |
GenBank mRNA | AF140501.1 | - | 6 - 7 | 964 | 178 |
AF245438.1 | - | 6 - 7 | 935 | 178 | |
AK226057.1 | - | 4 - 5 | 602 | 178 | |
AK301578.1 | - | 5 - 6 | 763 | 178 | |
AL136670.1 | - | 6 - 7 | 949 | 178 | |
BC032617.1 | - | 5 - 6 | 1208 | 178 | |
BC032662.1 | - | 6 - 7 | 965 | 178 |