The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005169.3 | NP_005160.2 | 1 - 2 | 392 | 85 |
GenBank mRNA | AK290645.1 | - | 1 - 2 | 392 | 85 |
BC041564.1 | - | 1 - 2 | 374 | 85 | |
BQ890461.1 | - | 1 - 2 | 376 | 85 |