Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
Chromosome Location
Chr.11: 65061009 - 65084143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080668.3 NP_542399.1 1 - 2 207 64
XM_005273733.3 XP_005273790.1 1 - 2 202 64
XM_005273734.3 XP_005273791.1 1 - 2 202 64
XM_011544743.2 XP_011543045.1 1 - 2 202 64
XM_011544747.2 XP_011543049.1 1 - 2 193 64
XM_011544748.2 XP_011543050.1 1 - 2 202 64
XM_011544749.2 XP_011543051.1 1 - 2 202 64
XM_017017158.1 XP_016872647.1 1 - 2 202 64
GenBank mRNA AK291490.1 - 1 - 2 131 64
BC011000.2 - 1 - 2 115 64
CN332525.1 - 1 - 2 206 64

View Full Product Details