The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006239.2 | NP_006230.2 | 14 - 15 | 2112 | 94 |
XM_011532039.2 | XP_011530341.1 | 13 - 14 | 2345 | 94 | |
GenBank mRNA | AF023456.1 | - | 14 - 15 | 2122 | 94 |