The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014423.3 | NP_055238.1 | 18 - 19 | 3504 | 72 |
XM_005271963.4 | XP_005272020.1 | 19 - 20 | 3568 | 72 | |
XM_005271964.4 | XP_005272021.1 | 12 - 13 | 2216 | 72 | |
XM_006714587.3 | XP_006714650.1 | 17 - 18 | 3417 | 72 | |
GenBank mRNA | AF197927.1 | - | 18 - 19 | 3433 | 72 |
AF213987.2 | - | 18 - 19 | 3161 | 72 | |
BC137226.1 | - | 18 - 19 | 3402 | 72 |