The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014423.3 | NP_055238.1 | 6 - 7 | 1485 | 82 |
XM_005271963.4 | XP_005272020.1 | 7 - 8 | 1549 | 82 | |
GenBank mRNA | AB209547.1 | - | 6 - 7 | 1441 | 82 |
AF197927.1 | - | 6 - 7 | 1414 | 82 | |
AK024831.1 | - | 4 - 5 | 943 | 82 | |
AK055638.1 | - | 6 - 7 | 1433 | 82 | |
AK307407.1 | - | 6 - 7 | 1485 | 82 | |
BC137226.1 | - | 6 - 7 | 1383 | 82 | |
BC144538.1 | - | 7 - 8 | 1400 | 82 |